Monocaprylin ≥98.0% (por GC)
Proveedor: TCI
M1071-1G
M1071-5G
TCIAM1071-1GEA
97
EUR
TCIAM1071-1G
TCIAM1071-5G
Monocaprylin ≥98.0% (por GC)
Monocaprylin
Formula:
C₁₁H₂₂O₄ |
Número MDL:
MFCD00056652 Núm. CAS: 26402-26-6 EINECS: 247-668-1 Índice Merck: 14,06252 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...