5-Nitro-2-pyridinethiol ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
2-Mercapto-5-nitropyridine, 5-Nitro-2-pyridyl Mercaptan, 5-Nitro-2-pyridinethiol
M1102-5G
M1102-25G
TCIAM1102-5GEA
52
EUR
TCIAM1102-5G
TCIAM1102-25G
5-Nitro-2-pyridinethiol ≥98.0% (by titrimetric analysis)
5-Nitro-2-pyridinethiol
Formula:
C₅H₄N₂O₂S MW: 156,16 g/mol |
Número MDL:
MFCD00030890 Núm. CAS: 2127-09-5 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...