4-Methoxyphenylphosphonic acid ≥95.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
4-Phosphonylanisole
M1155-1G
M1155-250MG
TCIAM1155-1GEA
255
EUR
TCIAM1155-1G
TCIAM1155-250MG
4-Methoxyphenylphosphonic acid ≥95.0% (by titrimetric analysis)
4-Methoxyphenylphosphonic acid
Formula:
C₇H₉O₄P MW: 188,12 g/mol |
Número MDL:
MFCD00013948 Núm. CAS: 21778-19-8 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...