3-Sulphopropyl methacrylate potassium salt ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Methacrylic acid 3-sulphopropyl Ester Potassium salt, 3-sulphopropyl Methacrylate Potassium Salt, Potassium 3-(Methacryloyloxy)propanesulphonate

M1158-25G M1158-500G
TCIAM1158-25GEA 38 EUR
TCIAM1158-25G TCIAM1158-500G
3-Sulphopropyl methacrylate potassium salt ≥97.0% (by HPLC, titration analysis)
3-Sulphopropyl methacrylate potassium salt
Formula: C₇H₁₂O₅S
MW: 246.32 g/mol
Número MDL: MFCD00010139
Núm. CAS: 31098-21-2
EINECS: 250-466-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Bromination Method) min. 97.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR