2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol] ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: Bis[3-(benzotriazol-2-yl)-2-hydroxy-5-tert-octylphenyl]methane

M1259-25G M1259-500G
TCIAM1259-25GEA 35 EUR
TCIAM1259-25G TCIAM1259-500G
2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol] ≥98.0% (by HPLC)
2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol]
Formula: C₄₁H₅₀N₆O₂
MW: 658,89 g/mol
Número MDL: MFCD00192289
Núm. CAS: 103597-45-1
EINECS: 403-800-1
Índice Merck: 14,01294
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65be0315a292f68ae36f035b13316a9b62a0b0a5&vid=85d5e4c80eb3daf61b95bd53e8d3f6ca9cec2769&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 195.0 to 199.0 deg-C
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR