Benzyl-D-(-)-mandelate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: D-(-)-Mandelic Acid Benzyl Ester, (R)-Benzyl 2-hydroxy-2-phenylacetate

M1354-25G M1354-5G
TCIAM1354-25GEA 77 EUR
TCIAM1354-25G TCIAM1354-5G
Benzyl-D-(-)-mandelate ≥98.0% (por GC)
Benzyl-D-(-)-mandelate
Formula: C₁₅H₁₄O₃
MW: 242,27 g/mol
Número MDL: MFCD00674031
Núm. CAS: 97415-09-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 105.0 to 109.0 deg-C
Specific rotation [a]20/D -31.0 to -37.0 deg (C=1, CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR