Metrizoic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

M1363-25G M1363-5G
TCIAM1363-25GEA 135 EUR
TCIAM1363-25G TCIAM1363-5G
Metrizoic acid ≥98.0% (by HPLC, titration analysis)
Metrizoic acid
Formula: C₁₂H₁₁I₃N₂O₄
Número MDL: MFCD00867964
Núm. CAS: 1949-45-7
EINECS: 217-761-1
Índice Merck: 14,06156
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3fd42cabfe1653f0472acf874d7ffc0cb4a00d3&vid=b3fd42cabfe1653f0472acf874d7ffc0cb4a00d3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR