2-(Methylamino)isobutyric acid hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: N,2-Dimethylalanine

M1388-1G M1388-5G
TCIAM1388-1GEA 97 EUR
TCIAM1388-1G TCIAM1388-5G
2-(Methylamino)isobutyric acid hydrate ≥98.0% (by titrimetric analysis)
2-(Methylamino)isobutyric acid hydrate
Formula: C₅H₁₁NO₂
MW: 117,15 g/mol
Número MDL: MFCD00004191
Núm. CAS: 2566-34-9
EINECS: 219-898-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR