Methyl-2,3,4-tri-O-benzoyl-ɑ-D-glucopyranoside ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triyl tribenzoate, Methyl 2,3,4-Tri-O-benzoyl-α-D-glucopyranoside
M1487-1G
M1487-5G
TCIAM1487-1GEA
239
EUR
TCIAM1487-1G
TCIAM1487-5G
Methyl-2,3,4-tri-O-benzoyl-ɑ-D-glucopyranoside ≥98.0% (by HPLC)
Methyl-2,3,4-tri-O-benzoyl-alpha-D-glucopyranoside
Formula:
C₂₈H₂₆O₉ MW: 506,51 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD06797128 Núm. CAS: 34234-44-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 144.0 to 148.0 deg-C |
Specific rotation [a]20/D | +52.0 to +55.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...