4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
Proveedor: TCI
M1589-1G
M1589-5G
TCIAM1589-1GEA
82
EUR
TCIAM1589-1G
TCIAM1589-5G
4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-allyl-4,6-O-benzylidene-beta-D-galactopyranoside
Formula:
C₂₃H₂₆O₇ MW: 414.45 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD06797132 Núm. CAS: 400091-05-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 222.0 to 228.0 deg-C |
Specific rotation [a]20/D | -45.0 to -50.0 deg (C=0.5,pyridine) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...