4-(4-Methylpiperazin-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)

Proveedor: TCI

M1624-25G M1624-5G
TCIAM1624-25GEA 191 EUR
TCIAM1624-25G TCIAM1624-5G
4-(4-Methylpiperazin-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)
4-(4-Methylpiperazin-1-yl)benzaldehyde
Formula: C₁₂H₁₆N₂O
MW: 204,2682 g/mol
Número MDL: MFCD00452233
Núm. CAS: 27913-99-1

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 62.0 to 67.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR