Mono-6-O-(p-toluenesulphonyl)-ɑ-cyclodextrin ≥85.0% (by HPLC)
Proveedor: TCI
M1644-200MG
M1644-1G
TCIAM1644-200MGEA
245
EUR
TCIAM1644-200MG
TCIAM1644-1G
Mono-6-O-(p-toluenesulphonyl)-ɑ-cyclodextrin ≥85.0% (by HPLC)
Mono-6-O-(p-toluenesulphonyl)-alpha-cyclodextrin
Formula:
C₄₃H₆₆O₃₂S MW: 1127.03 g/mol Temperatura de almacenaje: Congelador |
Núm. CAS:
32860-56-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 85.0 area% |
Specific rotation [a]20/D | +117 to +127 deg (C=4, DMSO) (calcd.on anh.substance) |
Water | max. 15.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...