Milrinone ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

Sinónimos: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile, 2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile, 3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone

M1663-1G M1663-100MG
TCIAM1663-1GEA 187 EUR
TCIAM1663-1G TCIAM1663-100MG
Milrinone ≥98.0% (by HPLC, total nitrogen)
Milrinone
Formula: C₁₂H₉N₃O
MW: 211,22 g/mol
Número MDL: MFCD00133539
Núm. CAS: 78415-72-2
EINECS: 278-903-6
UN: 2811
ADR: 6.1,III
Índice Merck: 14,06197

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR