Miconazole nitrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate

M1769-25G M1769-5G
TCIAM1769-25GEA 270 EUR
TCIAM1769-25G TCIAM1769-5G
Miconazole nitrate ≥98.0% (by titrimetric analysis)
Miconazole nitrate
Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
Número MDL: MFCD00058161
Núm. CAS: 22832-87-7
EINECS: 245-256-6
Índice Merck: 14,06178

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR