2-(2-Methoxyphenoxy)ethanamine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 1-(2-Aminoethoxy)-2-methoxybenzene hydrochloride, Carvedilol EP IMpurity E, 2-(2-Methoxyphenoxy)ethylamine hydrochloride hydrate, 2-(2-Aminoethoxy)anisole hydrochloride

M1794-25G M1794-5G
TCIAM1794-25GEA 45 EUR
TCIAM1794-25G TCIAM1794-5G
2-(2-Methoxyphenoxy)ethanamine hydrochloride ≥98.0% (by titrimetric analysis)
2-(2-Methoxyphenoxy)ethanamine hydrochloride
Formula: C₉H₁₄ClNO₂
MW: 203,6660 g/mol
Número MDL: MFCD01631120
Núm. CAS: 64464-07-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR