17alpha-Methylandrostan-17beta-ol-3-one ≥93.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
17beta-Hydroxy-17alpha-methyl-5alpha-androstan-3-one, Mestanolone, Methylandrostanolone, Mestaline
M1801-5G
M1801-1G
TCIAM1801-5GEA
234
EUR
TCIAM1801-5G
TCIAM1801-1G
17alpha-Methylandrostan-17beta-ol-3-one ≥93.0% (by HPLC)
17alpha-Methylandrostan-17beta-ol-3-one
Formula:
C₂₀H₃₂O₂ MW: 304.47 g/mol |
Número MDL:
MFCD00065900 Núm. CAS: 521-11-9 EINECS: 208-302-6 Índice Merck: 14,05915 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Melting point | 189.0 to 193.0 deg-C |
Specific rotation [a]20/D | +9.0 to +11.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...