1-Methyl-L-aspartate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

M1859-1G M1859-5G
TCIAM1859-1GEA 16 EUR
TCIAM1859-1G TCIAM1859-5G
1-Methyl-L-aspartate ≥98.0% (by titrimetric analysis)
1-Methyl-L-aspartate
Formula: C₅H₉NO₄
Pto. de fusión: 186°C
Número MDL: MFCD00080289
Núm. CAS: 17812-32-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Specific rotation (value) 33.0 to 38.0 deg
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +33.0 to +38.0 deg (C=0.5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR