Methyl-4-Chloro-3-methylbenzoate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
4-Chloro-3-methylbenzoic Acid Methyl Ester, Methyl 4-chloro-3-methylbenzoate
M1935-25G
M1935-5G
TCIAM1935-25GEA
122
EUR
TCIAM1935-25G
TCIAM1935-5G
Methyl-4-Chloro-3-methylbenzoate ≥98.0% (por GC)
Methyl-4-Chloro-3-methylbenzoate
Formula:
C₉H₉ClO₂ MW: 184,62 g/mol |
Número MDL:
MFCD00151838 Núm. CAS: 91367-05-4 |
Resultados test especificaciones
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...