N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI

M1971-25G M1971-100G
TCIAM1971-25GEA 27 EUR
TCIAM1971-25G TCIAM1971-100G
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine ≥98.0% (by total nitrogen and titration analysis)
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine
Formula: C₁₁H₂₁NO₅S
Pto. de fusión: 149°C
Número MDL: MFCD00040574
Núm. CAS: 3637-26-1
EINECS: 222-860-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR