Methyl-4-(aminomethyl)cyclohexanecarboxylate hydrochloride (cis and trans mixture) cis- and trans-mixture ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride, Methyl 4-(aminomethyl)cyclohexanecarboxylate hydrochloride
M1987-1G
M1987-5G
TCIAM1987-1GEA
78
EUR
TCIAM1987-1G
TCIAM1987-5G
Methyl-4-(aminomethyl)cyclohexanecarboxylate hydrochloride (cis and trans mixture) cis- and trans-mixture ≥98.0% (by titrimetric analysis)
Methyl-4-(aminomethyl)cyclohexanecarboxylate hydrochloride (cis and trans mixture)
Formula:
C₉H₁₇NO₂ MW: 207.70 g/mol |
Número MDL:
MFCD03093748 Núm. CAS: 54640-02-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...