Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
Methyl trans-4-(aminomethyl)cyclohexane-1-carboxylate hydrochloride, trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
M1988-25G
M1988-5G
TCIAM1988-25GEA
453
EUR
TCIAM1988-25G
TCIAM1988-5G
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride ≥98.0% (by GC, titration analysis)
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride
Formula:
C₉H₁₇NO₂ MW: 207,70 g/mol |
Número MDL:
MFCD00797465 Núm. CAS: 29275-88-5 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...