(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride ≥95.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
H-Asp(OMe)-OH·HCl, L-Aspartic acid 4-methyl ester hydrochloride, L-Aspartatic Acid 4-Methyl Ester Hydrochloride, L-Aspartic acid β-methyl ester hydrochloride, H-Asp(OMe)-OH.HCl, H-Asp(OMe)-OH β-Methyl L-aspartate hydrochloride
M2024-25G
M2024-5G
TCIAM2024-25GEA
202
EUR
TCIAM2024-25G
TCIAM2024-5G
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride ≥95.0% (by titrimetric analysis)
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
Formula:
C₅H₉NO₄·HCl MW: 183,6 g/mol Pto. de fusión: 193°C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00038972 Núm. CAS: 16856-13-6 EINECS: 240-880-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Specific rotation [a]20/D | +9.0 to +13.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...