Methyl-2,3:4,6-di-O-benzylidene-ɑ-D-mannopyranoside ≥98.0% (by HPLC)

Proveedor: TCI

M2061-25G M2061-5G
TCIAM2061-25GEA 359 EUR
TCIAM2061-25G TCIAM2061-5G
Methyl-2,3:4,6-di-O-benzylidene-ɑ-D-mannopyranoside ≥98.0% (by HPLC)
Methyl-2,3:4,6-di-O-benzylidene-alpha-D-mannopyranoside
Formula: C₂₁H₂₂O₆
MW: 370,40 g/mol
Número MDL: MFCD00192286
Núm. CAS: 4148-71-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 183.0 to 187.0 deg-C
Specific rotation [a]20/D -1.0 to +1.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR