Methocarbamol ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
M2254-25G
M2254-5G
TCIAM2254-25GEA
77
EUR
TCIAM2254-25G
TCIAM2254-5G
Methocarbamol ≥98.0% (by HPLC, total nitrogen)
Methocarbamol
Formula:
C₁₁H₁₅NO₅ |
Número MDL:
MFCD00057662 Núm. CAS: 532-03-6 EINECS: 208-524-3 Índice Merck: 14,05980 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 94.0 to 98.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...