DL-4-Methoxymandelic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Hydroxy-2-(4-methoxyphenyl)acetic acid

M2278-5G M2278-25G
TCIAM2278-5GEA 65 EUR
TCIAM2278-5G TCIAM2278-25G
DL-4-Methoxymandelic acid ≥98.0% (by HPLC, titration analysis)
DL-4-Methoxymandelic acid
Formula: C₉H₁₀O₄
MW: 182,18 g/mol
Número MDL: MFCD00004233
Núm. CAS: 10502-44-0
EINECS: 234-031-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94f0cb9afa8232153b2023a6b92b4e5457dfa94d&vid=94f0cb9afa8232153b2023a6b92b4e5457dfa94d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR