Monosodium risedronate hemipentahydrate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Monosodium 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonate, Risedronic Acid Monosodium Salt
M2289-100MG
M2289-1G
TCIAM2289-100MGEA
73
EUR
TCIAM2289-100MG
TCIAM2289-1G
Monosodium risedronate hemipentahydrate ≥98.0% (by HPLC, titration analysis)
Monosodium risedronate hemipentahydrate
Formula:
C₇H₁₁NO₇P₂ MW: 305.09 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD01706268 Núm. CAS: 329003-65-8 Índice Merck: 14,08232 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 11.9 to 13.9 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...