Methyl-2-chloro-D-mandelate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Methyl (R)-2'-Chloro-alpha-hydroxyphenylacetate, (R)-Methyl 2-(2-chlorophenyl)-2-hydroxyacetate, 2-Chloro-D-mandelic Acid Methyl Ester, (R)-2'-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester
M2382-5G
M2382-25G
TCIAM2382-5GEA
68
EUR
TCIAM2382-5G
TCIAM2382-25G
Methyl-2-chloro-D-mandelate ≥98.0% (por GC)
Methyl-2-chloro-D-mandelate
Formula:
C₉H₉ClO₃ MW: 200,62 g/mol |
Número MDL:
MFCD02259751 Núm. CAS: 32345-59-8 |
Resultados test especificaciones
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -178 to -182 deg (C=0.4, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...