Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) ≥96.0% (por GC)
Proveedor: TCI
Sinónimos:
Methyl 3-oxo-4-phenylbutanoate, Methyl 4-Phenylacetoacetate, 3-Oxo-4-phenylbutyric Acid Methyl Ester, 4-Phenylacetoacetic Acid Methyl Ester
M2396-1G
M2396-5G
TCIAM2396-1GEA
38
EUR
TCIAM2396-1G
TCIAM2396-5G
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) ≥96.0% (por GC)
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers)
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Pto. Ebullición: 136°C |
Número MDL:
MFCD03424746 Núm. CAS: 37779-49-0 |
|
Resultados test especificaciones
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 96.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...