4-Methyl-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
M2457-1G
M2457-5G
TCIAM2457-1GEA
30
EUR
TCIAM2457-1G
TCIAM2457-5G
4-Methyl-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Methyl-1-naphthaleneboronic acid
Formula:
C₁₁H₁₁BO₂ MW: 186,02 g/mol Pto. de fusión: 207°C |
Número MDL:
MFCD01632204 Núm. CAS: 103986-53-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 and le 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...