(3-Methoxybenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

M2463-1G M2463-5G
TCIAM2463-1GEA 49 EUR
TCIAM2463-1G TCIAM2463-5G
(3-Methoxybenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(3-Methoxybenzyl)triphenylphosphonium chloride
Formula: C₂₆H₂₄ClOP
MW: 418,90 g/mol
Número MDL: MFCD09836189
Núm. CAS: 18880-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8f6f6206307efc459c45e1b3a082ba8b3c2ef9&vid=8a8f6f6206307efc459c45e1b3a082ba8b3c2ef9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR