Moxifloxacin hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Moxifloxacin HCl monohydrate, 1-Cyclopropyl-6-fluoro-7-((4aS,7aS)-hexahydro-1H-pyrrolo[3,4-b]pyridin-6(2H)-yl)-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride hydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid hydrochloride hydrate
M2479-100MG
M2479-1G
TCIAM2479-100MGEA
77
EUR
TCIAM2479-100MG
TCIAM2479-1G
Moxifloxacin hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)
Moxifloxacin hydrochloride monohydrate
Formula:
C₂₁H₂₄FN₃O₄·HCl·H₂O MW: 455,91 g/mol |
Número MDL:
MFCD00949117 Núm. CAS: 192927-63-2 Índice Merck: 14,06291 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -123.5 to -136.5 deg (C=0.5, water) |
Water | 3.0 to 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...