Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Nicardipine USP Related Compound A, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid Monomethyl ester, 5-(Methoxycarbonyl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid
M2493-5G
M2493-25G
TCIAM2493-5GEA
29
EUR
TCIAM2493-5G
TCIAM2493-25G
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
Formula:
C₁₆H₁₆N₂O₆ MW: 332,31 g/mol |
Número MDL:
MFCD00475478 Núm. CAS: 74936-72-4 EINECS: 278-034-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...