1-Methyl-1H-pyrazole-5-carboxylic acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-Methyl-5-pyrazolecarboxylic acid

M2494-1G M2494-5G
TCIAM2494-1GEA 47 EUR
TCIAM2494-1G TCIAM2494-5G
1-Methyl-1H-pyrazole-5-carboxylic acid ≥97.0% (by GC, titration analysis)
1-Methyl-1H-pyrazole-5-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126,11 g/mol
Número MDL: MFCD00464253
Núm. CAS: 16034-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c34c88a7e379407a44b05951bfcabf12b5d54eeb&vid=c34c88a7e379407a44b05951bfcabf12b5d54eeb&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR