Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, 1-Benzyl-4-oxo-3-piperidinecarboxylic acid methyll ester hydrochloride, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate HCl, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
M2531-5G
M2531-25G
TCIAM2531-5GEA
20
EUR
TCIAM2531-5G
TCIAM2531-25G
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride ≥98.0% (by HPLC)
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Formula:
C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl MW: 283,75 g/mol |
Número MDL:
MFCD00012799 Núm. CAS: 3939-01-3 EINECS: 223-522-2 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...