Methyl-2-(3-fluorophenyl)acetate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Methyl 2-(3-fluorophenyl)acetate, Methyl 3-Fluorophenylacetate, 3-Fluorophenylacetic acid methyll ester, Methyl 3-fluorophenylacetate
M2536-5G
M2536-25G
TCIAM2536-5GEA
56
EUR
TCIAM2536-5G
TCIAM2536-25G
Methyl-2-(3-fluorophenyl)acetate ≥98.0% (por GC)
Methyl-2-(3-fluorophenyl)acetate
Formula:
C₉H₉FO₂ MW: 168.167 g/mol |
Número MDL:
MFCD03093288 Núm. CAS: 64123-77-9 |
|
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...