Mepivacaine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

M2560-1G M2560-5G
TCIAM2560-1GEA 87 EUR
TCIAM2560-1G TCIAM2560-5G
Mepivacaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Mepivacaine hydrochloride
Formula: C₁₅H₂₂N₂O
Número MDL: MFCD00144738
Núm. CAS: 1722-62-9
EINECS: 217-023-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f73e46a29435473c474dc0ae08fd6c292dbc120d&vid=f73e46a29435473c474dc0ae08fd6c292dbc120d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR