1-Methyl-4-(methylamino)piperidine ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
M2566-200MG
M2566-1G
TCIAM2566-200MGEA
22
EUR
TCIAM2566-200MG
TCIAM2566-1G
1-Methyl-4-(methylamino)piperidine ≥97.0% (by GC, titration analysis)
1-Methyl-4-(methylamino)piperidine
Formula:
C₇H₁₆N₂ Pto. Ebullición: 53°C |
Número MDL:
MFCD00006498 Núm. CAS: 73579-08-5 EINECS: 277-542-1 UN: 2920 ADR: 8,II |
|
Resultados test especificaciones
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...