Methyl-2-aminothiazole-4-carboxylate ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: Methyl 2-aminothiazole-4-carboxylate

M2581-1G M2581-5G
TCIAM2581-1GEA 37 EUR
TCIAM2581-1G TCIAM2581-5G
Methyl-2-aminothiazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Methyl-2-aminothiazole-4-carboxylate
Formula: C₅H₆N₂O₂S
MW: 158,1800 g/mol
Número MDL: MFCD00622441
Núm. CAS: 118452-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4b0d6822fb7a03ee69833a562b1bfd15a0d797d&vid=d4b0d6822fb7a03ee69833a562b1bfd15a0d797d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 171.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR