2-(4-Methyl-1-piperazinyl)aniline ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 1-(2-Aminophenyl)-4-methylpiperazine

M2592-1G M2592-5G
TCIAM2592-1GEA 35 EUR
TCIAM2592-1G TCIAM2592-5G
2-(4-Methyl-1-piperazinyl)aniline ≥98.0% (by GC, titration analysis)
2-(4-Methyl-1-piperazinyl)aniline
Formula: C₁₁H₁₇N₃
MW: 191,28 g/mol
Número MDL: MFCD04035359
Núm. CAS: 180605-36-1
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fe9e40d7bacd9dfb191bedebf7eaf0ce8e1d075&vid=9fe9e40d7bacd9dfb191bedebf7eaf0ce8e1d075&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR