1-(4-Metoxifenil)-2-tiourea ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Peligro
|
M2593-5G
M2593-25G
TCIAM2593-5GEA
95
EUR
TCIAM2593-5G
TCIAM2593-25G
1-(4-Metoxifenil)-2-tiourea ≥98.0% (by HPLC, total nitrogen)
1-(4-Metoxifenil)-2-tiourea
Formula:
(C₆H₄(OCH₃))NC(S)NH₂ |
Número MDL:
MFCD00004936 Núm. CAS: 2293-07-4 EINECS: 218-931-8 UN: 2811 ADR: 6.1,III |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 206.0 to 210.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...