Methyl-5-bromopicolinate ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate
M2614-1G
M2614-5G
TCIAM2614-1GEA
29
EUR
TCIAM2614-1G
TCIAM2614-5G
Methyl-5-bromopicolinate ≥98.0% (por GC)
Methyl-5-bromopicolinate
Formula:
C₇H₆BrNO₂ MW: 216,03 g/mol |
Número MDL:
MFCD04112493 Núm. CAS: 29682-15-3 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...