Methyl-2-bromothiazole-4-carboxylate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Methyl 2-bromothiazole-4-carboxylate
M2615-1G
M2615-5G
TCIAM2615-1GEA
36
EUR
TCIAM2615-1G
TCIAM2615-5G
Methyl-2-bromothiazole-4-carboxylate ≥98.0% (por GC)
Methyl-2-bromothiazole-4-carboxylate
Formula:
C₅H₄BrNO₂S MW: 222,06 g/mol |
Número MDL:
MFCD06659908 Núm. CAS: 170235-26-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...