trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl ≥98.0% (por GC)

Proveedor: TCI

M2638-1G M2638-5G
TCIAM2638-1GEA 60 EUR
TCIAM2638-1G TCIAM2638-5G
trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl ≥98.0% (por GC)
trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl
Formula: C₂₁H₃₀
Número MDL: MFCD12911573
Núm. CAS: 155041-85-3
EINECS: 439-730-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0232911221c936983e20e7612e66fddca0744af0&vid=0232911221c936983e20e7612e66fddca0744af0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR