Moxonidine ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: 4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine, 4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methylpyrimidin-5-amine

M2660-200MG M2660-1G
TCIAM2660-200MGEA 59 EUR
TCIAM2660-200MG TCIAM2660-1G
Moxonidine ≥98.0% (by titrimetric analysis)
Moxonidine
Formula: C₉H₁₂ClN₅O
MW: 241,68 g/mol
Número MDL: MFCD00865794
Núm. CAS: 75438-57-2
UN: 2811
ADR: 6.1,III
Índice Merck: 14,06293
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75c546bc092367c7b0a93cd0bd5baebf716a289a&vid=75c546bc092367c7b0a93cd0bd5baebf716a289a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR