3-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(3-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxycarbonylphenylboronic acid pinacol ester
M2665-1G
M2665-5G
TCIAM2665-1GEA
38
EUR
TCIAM2665-1G
TCIAM2665-5G
3-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₄H₁₉BO₄ MW: 262,1093 g/mol |
Número MDL:
MFCD03789258 Núm. CAS: 480425-35-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...