3-Methacrylamidophenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
M2676-1G
M2676-5G
TCIAM2676-1GEA
52
EUR
TCIAM2676-1G
TCIAM2676-5G
3-Methacrylamidophenylboronic acid (contains varying amounts of Anhydride)
3-Methacrylamidophenylboronic acid
Formula:
C₁₀H₁₂BNO₃ |
Número MDL:
MFCD09475805 Núm. CAS: 48150-45-4 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...