Z-Asp-OMe ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 1-Methyl N-Benzyloxycarbonyl-L-aspartate, N-Benzyloxycarbonyl-L-aspartic acid 1-Methyl ester, Cbz-Asp-OMe, 1-Methyl N-Cbz-L-aspartate, Z-Asp-OMe, N-Cbz-L-aspartic acid 1-Methyl ester, N-Benzyloxycarbonyl-L-aspartic acid 1-methyl ester, (S)-3-(((Benzyloxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid, 1-Methyl N-Carbobenzoxy-L-aspartate, N-Carbobenzoxy-L-aspartic acid 1-Methyl ester

M2707-5G M2707-25G
TCIAM2707-5GEA 64 EUR
TCIAM2707-5G TCIAM2707-25G
Z-Asp-OMe ≥98.0% (by HPLC, titration analysis)
Z-Asp-OMe
Formula: C₁₃H₁₅NO₆
MW: 281.264 g/mol
Número MDL: MFCD00083277
Núm. CAS: 4668-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc1333d119bbb0e9da357c47195cce1b02c4a8d8&vid=cc1333d119bbb0e9da357c47195cce1b02c4a8d8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 89.0 to 93.0 deg-C
Specific rotation [a]20/D -15.0 to -18.0 deg (C=2,EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR