4-Metilfenilamidina clorhidrato ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Methylphenylamidine hydrochloride, 4-Methylbenzene-1-carboximidamide hydrochloride, 4-Methylbenzimidamide hydrochloride
M2714-1G
M2714-5G
TCIAM2714-1GEA
43
EUR
TCIAM2714-1G
TCIAM2714-5G
4-Metilfenilamidina clorhidrato ≥98.0% (by HPLC, total nitrogen)
4-Metilfenilamidina clorhidrato
Formula:
C₈H₁₁ClN₂ MW: 170.64 g/mol |
Número MDL:
MFCD00040183 Núm. CAS: 6326-27-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 213.0 to 217.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...