Methyl-1-boc-3-pyrrolidinecarboxylate ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Pyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, 1-Boc-3-pyrrolidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Pyrrolidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate, Methyl 1-Boc-3-pyrrolidinecarboxylate
M2723-1G
M2723-5G
TCIAM2723-1GEA
61
EUR
TCIAM2723-1G
TCIAM2723-5G
Methyl-1-boc-3-pyrrolidinecarboxylate ≥98.0% (por GC)
Methyl-1-boc-3-pyrrolidinecarboxylate
Formula:
C₁₁H₁₉NO₄ MW: 229,28 g/mol |
Número MDL:
MFCD04038683 Núm. CAS: 122684-33-7 |
Resultados test especificaciones
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...