4-Methylphenylalanine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
H-Phe(4-Me)-OH, H-p-Me-Phe-OH, 4-Methyl-L-phenylalanine
M2743-1G
M2743-5G
TCIAM2743-1GEA
60
EUR
TCIAM2743-1G
TCIAM2743-5G
4-Methylphenylalanine ≥98.0% (by HPLC, titration analysis)
4-Methylphenylalanine
Formula:
C₁₀H₁₃NO₂ MW: 179.219 g/mol |
Número MDL:
MFCD00038575 Núm. CAS: 1991-87-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -8.0 to -11.0 deg (C=1,1mol/L HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...